Learn More
Alkali Scientific CellPro™ WeighBucket, Weigh Boat, 40mL, with handle
SDP
Catalog No. 503027654
Encompass_Preferred
New polyethylene WeighBuckets™ are a new type of weigh boat that safely transports, contains and pours liquids, powders and small solids. The handle and spout make them easy to use, practically eliminating spillage and making controlled-rate pouring possible. Dimensions excluding rim, spout and handle: 53 x 33mm (D x H) Easy-grip handle 40mL capcity Convenient pour spot Packing: 500/pk
Catalog No. 50-302-7654 Supplier Alkali Scientific Supplier No. J6602W
New polyethylene WeighBuckets™ are a new type of weigh boat that safely transports, contains and pours liquids, powders and small solids. The handle and spout make them easy to use, practically eliminating spillage and making controlled-rate pouring possible. Dimensions excluding rim, spout and handle: 53 x 33mm (D x H) Easy-grip handle 40mL capcity Convenient pour spot Packing: 500/pk