Learn More
Alkali Scientific CellPro™ X-Gal Solution, 1X 40mg/mL 100mL
SDP
Catalog No. 502389302
Encompass_Preferred
CellPro™ X-Gal (5-bromo-4-chloro-3-indolyl-β-D-galactopyranoside) Solution stands as a trusted reagent in molecular biology research. Crafted to stringent standards of purity and sterility, this solution provides researchers with a reliable substrate for β-galactosidase assays. With its sterile filtered formulation, CellPro™ X-Gal Solution ensures consistent and reproducible results, empowering researchers in their molecular biology experiments.
Catalog No. 50-238-9302 Supplier Alkali Scientific Supplier No. DS0205
Add to Cart
Add to Cart
CellPro™ X-Gal (5-bromo-4-chloro-3-indolyl-β-D-galactopyranoside) Solution stands as a trusted reagent in molecular biology research. Crafted to stringent standards of purity and sterility, this solution provides researchers with a reliable substrate for β-galactosidase assays. With its sterile filtered formulation, CellPro™ X-Gal Solution ensures consistent and reproducible results, empowering researchers in their molecular biology experiments.