Learn More
Sterlitech Corporation CELLQART 12WELL CELL CULT 48PK
SDP
Catalog No. NC2314948
Encompass_Preferred
CellQART 12-Well Cell Culture Insert PET, 3.0um, Clear, 48/pk. Ease of use by convenient pipette access (patented design since 2011). Optimized gas exchange by product design. Compatible to standard well-plates. Industry leading quality standards to ensure membrane parameter consistency (important for consistent cell culture results). RNase/DNase Free and Non-pyrogenic. DIN EN ISO 9001 and DIN EN ISO 13485 certified
Catalog No. NC2314948 Supplier Sterlitech Corporation Supplier No. 9313012
CellQART 12-Well Cell Culture Insert PET, 3.0um, Clear, 48/pk. Ease of use by convenient pipette access (patented design since 2011). Optimized gas exchange by product design. Compatible to standard well-plates. Industry leading quality standards to ensure membrane parameter consistency (important for consistent cell culture results). RNase/DNase Free and Non-pyrogenic. DIN EN ISO 9001 and DIN EN ISO 13485 certified