Learn More
BINDER™ CELLROLL Roller Bottle System
SDP
Catalog No. 5000217494 Shop All BINDER Inc. Products
Click to view available options
For Use With (Equipment):
BINDER Units CB 220
Catalog No. For Use With (Equipment)
50-002-17494 BINDER Units CB 220
1 options
Catalog No. 50-002-17494 Supplier BINDER™ Supplier No. 80120572
Add to Cart
Add to Cart
Request a Quote
For Use With: BINDER units CB 220
  • Modular
  • Expandable
  • For either 4 or 6 roller bottles
Specifications
Description CELLROLL Set - modular, expandable roller bottle system for cell cultivation, 6 roller bottles
For Use With (Equipment) BINDER Units CB 220
Holds 6 roller bottles
Type Roller Bottle System
For Use With (Application) Cell cultivation