Learn More
BINDER™ CELLROLL Roller Bottle System
SDP
Catalog No. 5000217495 Shop All BINDER Inc. Products
Click to view available options
For Use With (Equipment):
CB 170
Catalog No. For Use With (Equipment)
50-002-17495 CB 170
1 options
Catalog No. 50-002-17495 Supplier BINDER™ Supplier No. 80121885
Add to Cart
Add to Cart
Request a Quote
For Use With: CB 170
  • Modular
  • Expandable roller bottle system for cell cultivation
  • 4 roller bottles
Specifications
Description CELLROLL set - modular, expandable roller bottle system for cell cultivation, 4 roller bottles
For Use With (Equipment) CB 170
Type Roller Bottle System
Includes 4 roller bottles