Learn More
Lifeline Cell Technology CELLS AIRWY EPI HU LIFELNE VL
SDP
Supplier:  Lifeline Cell Technology FC0016
Encompass_Preferred
Normal Human Airway Epithelial Cells, cultured without retinoic acid and cryopreserved as primary cells. Quality tested in BronchiaLife SAE Medium to ensure optimal growth over a period of at least 15 population doublings.
Catalog No. 50-311-853