Learn More
Lifeline Cell Technology CELLS AO ENDO HU LIFELNE VL
SDP
Catalog No. 50311851
Encompass_Preferred
Normal Human Aortic Endothelial Cells are cryopreserved, secondary. Quality tested in VascuLife 2% serum medium to ensure optimal reduced-serum growth. Not exposed to antimicrobials or phenol red.
Catalog No. 50-311-851 Supplier Lifeline Cell Technology Supplier No. FC0014
Add to Cart
Add to Cart
Normal Human Aortic Endothelial Cells are cryopreserved, secondary. Quality tested in VascuLife 2% serum medium to ensure optimal reduced-serum growth. Not exposed to antimicrobials or phenol red.