Learn More
Lifeline Cell Technology CELLS AO SMC HU LIFELNE VL
SDP
Catalog No. 50311852
Encompass_Preferred
Normal Human Aortic Smooth Muscle Cells (HAoSMC), when grown in Lifeline's VascuLife SMC Medium, provide an ideal low serum (5%) culture model for the study of angiogenesis, atherosclerosis, diabetes. Cryopreserved secondary cells.
Catalog No. 50-311-852 Supplier Lifeline Cell Technology Supplier No. FC0015
Add to Cart
Add to Cart
Normal Human Aortic Smooth Muscle Cells (HAoSMC), when grown in Lifeline's VascuLife SMC Medium, provide an ideal low serum (5%) culture model for the study of angiogenesis, atherosclerosis, diabetes. Cryopreserved secondary cells.