Learn More
Lifeline Cell Technology CELLS BLADR SM HU LIFELNE VL
SDP
Catalog No. 50311872
Encompass_Preferred
Human Bladder Smooth Muscle Cells, secondary, 500,000 cells per vial. Cells are sterility and virus tested. All tissues used for cell isolation are obtained under informed consent. Lifeline complies with the U.S. HIPAA.
Catalog No. 50-311-872 Supplier Lifeline Cell Technology Supplier No. FC0043
Human Bladder Smooth Muscle Cells, secondary, 500,000 cells per vial. Cells are sterility and virus tested. All tissues used for cell isolation are obtained under informed consent. Lifeline complies with the U.S. HIPAA.