Learn More
Lifeline Cell Technology Human Bronchial/Tracheal Epithelial Cells, Primary
SDP
Catalog No. 50311867
Encompass_Preferred
Lifeline normal Human Bronchial/Tracheal Epithelial Cells are cultured without retinoic acid, cryopreserved primary cells. Quality tested in BronchiaLife B/T Medium to ensure optimal growth over a period of at least 15 population doublings
Catalog No. 50-311-867 Supplier Lifeline Cell Technology Supplier No. FC0035
Lifeline normal Human Bronchial/Tracheal Epithelial Cells are cultured without retinoic acid, cryopreserved primary cells. Quality tested in BronchiaLife B/T Medium to ensure optimal growth over a period of at least 15 population doublings