Learn More
Lifeline Cell Technology CELLS CO ART SM HU LIFELNE VL
SDP
Catalog No. 50311864
Encompass_Preferred
Lifeline's normal Human Coronary Artery Smooth Muscle Cells (HCASMC), when grown in VascuLife SMC Medium, provide an ideal culture model for the study of angiogenesis, atherosclerosis, diabetes or vascular/pulmonary biology.
Catalog No. 50-311-864 Supplier Lifeline Cell Technology Supplier No. FC0031
Add to Cart
Add to Cart
Lifeline's normal Human Coronary Artery Smooth Muscle Cells (HCASMC), when grown in VascuLife SMC Medium, provide an ideal culture model for the study of angiogenesis, atherosclerosis, diabetes or vascular/pulmonary biology.