Learn More
LGC GENOMICS LLC (Lucigen) Endura Chemically Competent Cells, 12 Rxns (DUOs)Clone and maintain unstable or repetitive sequences and lentiviral libraries with high efficiency. Ideal for GeCKO CRISPR library transformation.
Catalog No. NC1886318
Encompass
Lucigens Endura Competent Cells are a commonly used strain for cloning sequences that suffer unwanted recombination events in other strains. Clones with inverted repeats or other sequences prone to recombination are commonly found in retroviral genes, and require cells such as Endura to be propagated stably.
Catalog No. NC1886318 Supplier LGC GENOMICS LLC Supplier No. 602401
Lucigens Endura Competent Cells are a commonly used strain for cloning sequences that suffer unwanted recombination events in other strains. Clones with inverted repeats or other sequences prone to recombination are commonly found in retroviral genes, and require cells such as Endura to be propagated stably.