Learn More
Lifeline Cell Technology CELLS CORNEL EPI HU LIFELNE VL
SDP
Supplier:  Lifeline Cell Technology FC0029
Encompass_Preferred
Normal Human Corneal Epithelial Cells (HCEC), when grown in Lifeline's OcuLife Medium, provide an ideal serum-free culture model for cell-matrix interactions, gene regulation and tissue development, drug development.
Catalog No. 50-311-862