Learn More
Lifeline Cell Technology CELLS FIBS HU X-FREE LIFELN VL
SDP
Catalog No. 50311868
Encompass_Preferred
Normal Human Dermal Fibroblasts-Neonatal, Xeno-Free, when grown in Lifeline's FibroLife Xeno-Free Complete Medium, provide an ideal xeno-free culture system to establish xeno-free human feeder layers for human embryonic stem cell cultures.
Catalog No. 50-311-868 Supplier Lifeline Cell Technology Supplier No. FC0037
Normal Human Dermal Fibroblasts-Neonatal, Xeno-Free, when grown in Lifeline's FibroLife Xeno-Free Complete Medium, provide an ideal xeno-free culture system to establish xeno-free human feeder layers for human embryonic stem cell cultures.