Learn More
Lifeline Cell Technology CELLS HUVEC LIFELINE VL
SDP
Supplier:  Lifeline Cell Technology FC0003
Encompass_Preferred
Normal Human Umbilical Vein Endothelial Cells (HUVEC), cryopreserved as primary cells. Quality tested in VascuLife 2% serum medium to ensure optimal reduced-serum growth over a period of at least 15 population doublings.
Catalog No. 50-311-847