Learn More
Lifeline Cell Technology CELLS HUVEC POOLD LIFELNE VL
SDP
Supplier:  Lifeline Cell Technology FC0044
Encompass_Preferred
HUVEC 10-Donor Pools, when grown in Lifeline's VascuLife Medium, provide an ideal low-serum culture model for the study of angiogenesis, atherosclerosis or vascular biology without the concern of atypical results from a single donor.
Catalog No. 50-311-873