Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
In this experiment, students will plan and execute their own investigation in which they will observe of a variety of samples under the microscope to explore the various parts of a cell and distinguish between living versus non-living substances, animal cells versus plant cells, and unicellular versus multicellular organisms.
Specifications
Specifications
| Grade | 6+ |
| Class Size | 15 Lab Groups |
| Product Type | School Microbiology Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
