Learn More
Lifeline Cell Technology CELLS KERATN HU AD LIFELNE VL
SDP
Supplier:  Lifeline Cell Technology FC0025
Encompass_Preferred
Lifeline's normal Human Epidermal Keratinocytes adult are cryopreserved as primary cells. Our HEK are quality tested in DermaLife K Medium to ensure optimal serum-free growth over a period of at least 15 population doublings.
Catalog No. 50-311-860