Learn More
Lifeline Cell Technology CELLS KERATN HU NEO LIFELNE VL
SDP
Supplier:  Lifeline Cell Technology FC0007
Encompass_Preferred
Normal Human Epidermal Keratinocytes neonatal are cryopreserved as primary cells. Our HEK are quality tested in DermaLife K Medium to ensure optimal serum-free growth over a period of at least 15 population doublings.
Catalog No. 50-311-848