Learn More
Lifeline Cell Technology CELLS KERATN HU POOL LIFELN VL
SDP
Supplier:  Lifeline Cell Technology FC0064
Encompass_Preferred
Normal Human Epidermal Keratinocytes 10-donor pool offers a wide range of donor demographics for research. Quality tested in DermaLife K Medium to ensure optimal serum-free growth over a period of at least 15 population doublings.
Catalog No. 50-311-883