Learn More
Lifeline Cell Technology CELLS LUNG FIBS HU LIFELN VL
SDP
Supplier:  Lifeline Cell Technology FC0049
Encompass_Preferred
Lifeline's normal Human Lung Fibroblasts provide an ideal cell system to establish serum free human feeder layers for human embryonic stem cell cultures or as a model to study wound healing, toxicology or basic cell biology.
Catalog No. 50-311-876