Learn More
Lifeline Cell Technology CELLS LUNG SM HU LD LIFELN VL
SDP
Catalog No. 50311874
Encompass_Preferred
Lifeline's normal Human Lung Smooth Muscle Cells, when grown in VascuLife SMC Medium, provide an ideal culture model for the study of angiogenesis, atherosclerosis, diabetes or vascular/pulmonary biology.
Catalog No. 50-311-874 Supplier Lifeline Cell Technology Supplier No. FC0046
Lifeline's normal Human Lung Smooth Muscle Cells, when grown in VascuLife SMC Medium, provide an ideal culture model for the study of angiogenesis, atherosclerosis, diabetes or vascular/pulmonary biology.