Learn More
Lifeline Cell Technology CELLS MELS HU AD LIFELNE VL
SDP
Catalog No. 50311863
Encompass_Preferred
Normal Human Epidermal Melanocytes adult are cryopreserved as secondary cells. Quality tested in DermaLife Ma Medium to ensure optimal reduced-serum growth over a period of at least 15 population doublings.
Catalog No. 50-311-863 Supplier Lifeline Cell Technology Supplier No. FC0030
Add to Cart
Add to Cart
Normal Human Epidermal Melanocytes adult are cryopreserved as secondary cells. Quality tested in DermaLife Ma Medium to ensure optimal reduced-serum growth over a period of at least 15 population doublings.