Learn More
Lifeline Cell Technology CELLS MICRO V HU NEO LIFELN VL
SDP
Catalog No. 50311871
Encompass_Preferred
Normal Human Dermal Microvascular Endothelial Cells (HD-MVECn), when grown in Lifeline's VascuLife Medium, provides an ideal culture model, with or without human VEGF, for the study of angiogenesis, atherosclerosis, or vascular biology.
Catalog No. 50-311-871 Supplier Lifeline Cell Technology Supplier No. FC0042
Normal Human Dermal Microvascular Endothelial Cells (HD-MVECn), when grown in Lifeline's VascuLife Medium, provides an ideal culture model, with or without human VEGF, for the study of angiogenesis, atherosclerosis, or vascular biology.