Learn More
Lifeline Cell Technology CELLS PROST EPI HU LIFELNE VL
SDP
Supplier:  Lifeline Cell Technology FC0038
Encompass_Preferred
Normal Human Prostate Epithelial Cells, cryopreserved as secondary cells for highest viability, plating efficiency. Quality tested in ProstaLife Medium to ensure optimal serum-free growth over a period of at least 15 population doublings.
Catalog No. 50-311-869