Learn More
Lifeline Cell Technology CELLS PUL ART END HU LIFELN VL
SDP
Catalog No. 50311880
Encompass_Preferred
Normal Human Pulmonary Artery Endothelial Cells (HPAEC), when grown in Lifeline's VascuLife Medium, provide an ideal low-serum culture model, with or without human VEGF, for the study of angiogenesis, atherosclerosis or vascular biology.
Catalog No. 50-311-880 Supplier Lifeline Cell Technology Supplier No. FC0055
Normal Human Pulmonary Artery Endothelial Cells (HPAEC), when grown in Lifeline's VascuLife Medium, provide an ideal low-serum culture model, with or without human VEGF, for the study of angiogenesis, atherosclerosis or vascular biology.