Learn More
Lifeline Cell Technology CELLS RENAL EPI HU LIFELNE VL
SDP
Catalog No. 50311849
Encompass_Preferred
Normal Human Renal Cortical Epithelial Cells are cryopreserved as primary cells. Quality tested in RenaLife Medium to ensure optimal reduced-serum growth over a period of at least 15 population doublings.
Catalog No. 50-311-849 Supplier Lifeline Cell Technology Supplier No. FC0012
Normal Human Renal Cortical Epithelial Cells are cryopreserved as primary cells. Quality tested in RenaLife Medium to ensure optimal reduced-serum growth over a period of at least 15 population doublings.