Learn More
Lifeline Cell Technology CELLS RENAL MDEPI HU LIFELN VL
SDP
Catalog No. 50311855
Encompass_Preferred
Normal Human Renal Medullary Epithelial Cells are cryopreserved as primary cells. Quality tested in RenaLife Medium to ensure optimal reduced-serum growth over a period of at least 15 population doublings.
Catalog No. 50-311-855 Supplier Lifeline Cell Technology Supplier No. FC0018
Normal Human Renal Medullary Epithelial Cells are cryopreserved as primary cells. Quality tested in RenaLife Medium to ensure optimal reduced-serum growth over a period of at least 15 population doublings.