Learn More
Electron Microscopy Sciences CellSafe™ Biopsy Capsule, Blue
SDP
Catalog No. 501931181
Encompass_Preferred
Eliminate artifacts by turning any cassette into a mesh cassette with these disposable plastic biopsy inserts have an integral nylon mesh and two hinged interlocking frames. With the extra fine mesh, you are ensured superior specimen security. Dimensions: 23L x 25W x 5mm H Mesh: 18L x 18W x 2mm H
Color: Blue
Catalog No. 50-193-1181 Supplier Electron Microscopy Sciences Supplier No. 62329
Eliminate artifacts by turning any cassette into a mesh cassette with these disposable plastic biopsy inserts have an integral nylon mesh and two hinged interlocking frames. With the extra fine mesh, you are ensured superior specimen security. Dimensions: 23L x 25W x 5mm H Mesh: 18L x 18W x 2mm H
Color: Blue