Learn More
Electron Microscopy Sciences CellSafe™ Biopsy Insert
SDP
Catalog No. 5029856
Encompass_Preferred
The CellSafe is a disposable plastic biopsy insert consisting of two hinged interlocking frames with an integral nylon mesh. The mesh is extra fine and therefore ensures a high level of specimen security. Measurements: Overall: 28 mm x 25 mm x 5 mm high (1.1" x 1" x 1/4"); The mesh area: 18 mm x 18 mm x 2 mm high.
Catalog No. 50-298-56 Supplier Electron Microscopy Sciences Supplier No. 6232710
The CellSafe is a disposable plastic biopsy insert consisting of two hinged interlocking frames with an integral nylon mesh. The mesh is extra fine and therefore ensures a high level of specimen security. Measurements: Overall: 28 mm x 25 mm x 5 mm high (1.1" x 1" x 1/4"); The mesh area: 18 mm x 18 mm x 2 mm high.