Learn More
SEXTON BIOTECHNOLOGIES INC 2 mL Clear Access Cryogenic Vials, 4/pack
SDP
Catalog No. NC1611396
Encompass
The difference is in its unique design. CellSeal cryogenic vials are created from USP class VI materials chosen for their chemical resistance, extreme durability under true cryogenic temperatures, and maximum cell recovery rates. With the patented CellSeal cryogenic vial design, contaminant risks are reduced and you gain a level of control, from early stage product concepts through commercialization, that your therapy and testing protocols demand.
Catalog No. NC1611396 Supplier SEXTON BIOTECHNOLOGIES INC Supplier No. CSV0514
The difference is in its unique design. CellSeal cryogenic vials are created from USP class VI materials chosen for their chemical resistance, extreme durability under true cryogenic temperatures, and maximum cell recovery rates. With the patented CellSeal cryogenic vial design, contaminant risks are reduced and you gain a level of control, from early stage product concepts through commercialization, that your therapy and testing protocols demand.