Learn More
Greiner Bio-One CELLSTAR 384-well, Cell Culture-Treated, Flat-Bottom Microplate
Catalog No. 07000879 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
07-000-879 Clear
1 options
Catalog No. 07-000-879 Supplier Greiner Bio-One Supplier No. 781182
Add to Cart
Edge
Add to Cart
Color:Clear; Sterility:Sterile; Lid:With Lid
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Non-cytotoxicCompatible with automated systemsFlexible working volume
  • Rounded square well profile
  • Stackable
  • Alphanumeric well cod
Service Plan
No
TRUSTED_SUSTAINABILITY
Specifications
Lid With Lid
Array 16 x 24
Color Clear
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 15 to 110 μL
Sterility Sterile
Surface Treatment Cell Culture
Total Volume (Metric) Well 131 μL
Well Shape Rounded Square
No. per Case 32
No. per Pack 8