Learn More
Greiner Bio-One CELLSTAR 384-well, Non-Binding, Flat-Bottom Microplate
GREENER_CHOICE
Catalog No. 07000900 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
07-000-900 Black
1 options
Catalog No. 07-000-900 Supplier Greiner Bio-One Supplier No. 781976
Add to Cart
Add to Cart
Color:Black; Sterility:Sterile; Lid:With Lid
  • Free of detectable DNase/RNase, human DNA and pyrogens
  • Non-cytotoxic
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Format Standard
Lid With Lid
Array 16 x 24
Autoclavable Not Autoclavable
Color Black
Product Type Cell Culture Microplate
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 15 to 110 μL
Sterility Sterile
Surface Treatment Non-binding
Total Volume (Metric) Well 131 μL
Well Shape Rounded Square
Dnase Free Yes
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 32
No. per Pack 1