Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One CELLSTAR 96-well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate
GREENER_CHOICE

Catalog No. 07000096 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-000-096 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-096 Supplier Greiner Bio-One Supplier No. 675083
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Color:White; Sterility:Sterile; Lid:With Lid

Alphanumeric coding
TRUSTED_SUSTAINABILITY

Specifications

Barcode Alphanumeric
Format Half-Area
Lid With Lid
Culture Area 0.15 cm2
Detection Method Luminescence
Array 8 x 12
Autoclavable Not Autoclavable
Color White
Product Type Cell Culture Microplate
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 15 to 175 μL
Sterility Sterile
Surface Treatment Tissue Culture
Total Volume (Metric) Well 199 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 32
No. per Pack 8
Quantity 32/Cs.
RNase Free Yes
Well Color White
Dimensions (L x W x H) 127.76 x 85.48 x 14.40 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.