Learn More
Greiner Bio-One CELLSTAR 96-well, Cell Culture-Treated, U-Shaped-Bottom Microplate
GREENER_CHOICE
Catalog No. 07000164 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
07-000-164 Clear
1 options
Catalog No. 07-000-164 Supplier Greiner Bio-One Supplier No. 650180
Add to Cart
Add to Cart
Color:Clear; Sterility:Sterile; Lid:With Lid
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Non-Cytotoxic
  • Compatible with automated systems
  • Alphanumeric well coding
  • Conforms to recommendation of American National Standards Institute (ANSI) (A
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Lid With Lid
Array 8 x 12
Autoclavable Not Autoclavable
Color Clear
Product Type Cell Culture Microplate
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 40 to 280 μL
Sterility Sterile
Surface Treatment Cell Culture
Total Volume (Metric) Well 323 μL
Well Shape Round Bottom (U-well)
Max. RCF 1000 x g (swinging-bucket rotor)
No. per Case 60
No. per Pack 1
Quantity 100/Cs.
Temperature Range (Metric) -20°C to +60°C