Learn More
Greiner Bio-One CELLSTAR 96-well, Cell Culture-Treated, V-Shaped-Bottom Microplate
GREENER_CHOICE
Catalog No. 07000080 Shop All Greiner Bio-One Products
Change view
Click to view available options
Lid:
Without Lid
Catalog No. Lid
07-000-080 Without Lid
1 options
Catalog No. 07-000-080 Supplier Greiner Bio-One Supplier No. 651160
Add to Cart
Add to Cart
Color:Clear; Sterility:Sterile; Lid:Without Lid
  • Compatible with automated systems
  • Alphanumeric well coding
  • Conforms to recommendation of American National Standards Institute (ANSI) (ANSI/SBS 1-2004)
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 8 x 12
Autoclavable Not Autoclavable
Color Clear
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 25 to 340 μL
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Conical Bottom (V-well)
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 75
No. per Pack 1
Quantity 100/Cs.
Temperature Range (Metric) -20°C to +60°C