Learn More
Greiner Bio-One CELLSTAR 96-well, Non-Treated, Flat-Bottom Microplate
GREENER_CHOICE
Catalog No. 07000108 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
07-000-108 Clear
1 options
Catalog No. 07-000-108 Supplier Greiner Bio-One Supplier No. 655185
Add to Cart
Add to Cart
Clear, sterile microplate with lid.
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Compatible with automated systems
  • Alphanumeric well coding
  • High clarity of microplates for optimal microscopic examinations
  • Stackable
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Format Standard
Lid With Lid
Array 8 x 12
Autoclavable Not Autoclavable
Color Clear
Product Type Cell Culture Microplate
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 25 to 340 μL
Sterility Sterile
Surface Treatment Untreated
Total Volume (Metric) Well 392 μL
Well Shape Flat Bottom (F-well)
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 60
No. per Pack 1
Quantity 60/Cs.
Temperature Range (Metric) -20°C to +60°C
Well Design Chimney