Learn More
Greiner Bio-One CELLSTAR 96-well, Non-Treated, U-Shaped-Bottom Microplate
GREENER_CHOICE
Catalog No. 07000133 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
07-000-133 Clear
1 options
Catalog No. 07-000-133 Supplier Greiner Bio-One Supplier No. 650185
Add to Cart
Add to Cart
Color:Clear; Sterility:Sterile; Lid:With Lid
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Non-Cytotoxic
  • Compatible with automated systems
  • Alphanumeric well coding
  • Conforms to recommendation of American National Standards Institute (ANSI)
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Format Standard
Lid With Lid
Array 8 x 12
Autoclavable Not Autoclavable
Color Clear
Product Type Cell Culture Microplate
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 40 to 280 μL
Sterility Sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 323 μL
Well Shape Round Bottom (U-well)
Max. RCF 1000 x g (swinging-bucket rotor)
No. per Case 60
No. per Pack 1
Temperature Range (Metric) -20°C to +60°C