Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One CELLSTAR Cell Culture Multi-well Plates for Suspension Cultures
GREENER_CHOICE

Catalog No. p-7075416 Shop All Greiner Bio-One Products
Change view
Click to view available options
Capacity (Metric):
1.7 mL
16 mL
3.3 mL (Well)
6.5 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
07-000-646 16 mL
07-000-670 3.3 mL (Well)
07-000-688 6.5 mL
07-000-727 1.7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07-000-646 Supplier Greiner Bio-One Supplier No. 657185
Only null left
Add to Cart
Add to Cart

Compatible with automated systems. Greiner Bio-One CELLSTAR Cell Culture Multi-well Plates for Suspension Cultures are used for anchorage dependant cell lines.

  • Manufactured from transparent virgin polystyrene
  • Low auto fluorescence
  • Single position lids to prevent cross-contamination
  • Alphanumeric well coding
  • With hydrophobic surface for suspension cultures and hybridoma cell

Service Plan

No

Warranty and Services

No

TRUSTED_SUSTAINABILITY

Specifications

Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Bottom Shape Flat
Capacity (Metric) 16 mL
Color Clear
Material Polystyrene
Shape Rectangular
Sterility Sterile
Surface Treatment Non-treated
Type Multi-well Cell Culture Plate
Well Count 6
Includes Lid
No. per Case 100
No. per Pack 1
Dimensions (L x W x H) 127.8 x 85.5 x16.5 mm
Length (Metric) 127.76 mm
Width (Metric) 85.48 mm
Height (Metric) 19 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.