Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One CELLSTAR Serological Pipette
GREENER_CHOICE

Catalog No. 07000788 Shop All Greiner Bio-One Products
Change view
Click to view available options
Quantity:
25/Pk., 1000/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-000-788 25/Pk., 1000/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-788 Supplier Greiner Bio-One Supplier No. 710107
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use in drop-free pipetting. Greiner Bio-One CELLSTAR Serological Pipettes are accurate with clear and legible graduation.

  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic, non-cytotoxic
  • Optical clarity
  • Accuracy
  • Pipette colour code according to international standards

Service Plan

No

Warranty and Services

No

TRUSTED_SUSTAINABILITY

Specifications

Aspirator Plug Yes
Autoclavable Not Autoclavable
Material Polystyrene
Sterility Sterile
For Use With (Application) Liquid Handling
Graduated Yes
Includes Sterile Serological Pipettes Packed In Plastic Bag
Quantity 25/Pk., 1000/Cs.
Tolerance ±2%
Packaging Type Bulk
Color Green
Volume (Metric) 2 mL
Type Pipette
Disposable Single-use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.