Learn More
Greiner Bio-One CELLSTAR Small Volume™ 384-well, Cell Culture-Treated, Flat-Bottom Microplate
Catalog No. 07000923 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
07-000-923 Black
1 options
Catalog No. 07-000-923 Supplier Greiner Bio-One Supplier No. 788086
Color:Black; Sterility:Sterile; Lid:With Lid
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic, non-cytotoxic
  • Compatible with automated systems
  • Stackable
  • Alphanumeric well coding
  • Growth area per well: 2.7mm²
  • Working volume per well:
TRUSTED_SUSTAINABILITY
Specifications
Lid With Lid
Description TC treated
Array 16 x 24
Color Black
Material Polystyrene
No. of Wells 384
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
No. per Pack 15/Pk.
Well Color Black
Well Design Chimney