Learn More
Greiner Bio-One CELLSTAR Small Volume™ 384-well, Flat-Bottom Microplate
Catalog No. 07000068 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
White
Catalog No. Color
07-000-068 White
1 options
Catalog No. 07-000-068 Supplier Greiner Bio-One Supplier No. 788075
Color:White; Sterility:Non-sterile; Lid:Without Lid
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic, non-cytotoxic
  • Compatible with automated systems
  • Stackable
  • Alphanumeric well coding
  • Growth area per well: 2.7mm²
  • Working volume per well:
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 16 x 24
Packaging 10/Pk., 80/Cs.
Binding Type Medium Binding
Color White
Product Type LoBase
Material Polystyrene
No. of Wells 384
Sterility Non-sterile
Well Shape Flat Bottom (F-well)
No. per Pack 10
Well Color White