Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One CELLSTAR Small Volume™ μClear™ 384-well, Cell Culture-Treated, Flat-Bottom Microplate

Catalog No. 07000035 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Black, Clear Bottom
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-000-035 Black, Clear Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-035 Supplier Greiner Bio-One Supplier No. 788091

missing translation for 'validMainframeMsg'

Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid

TRUSTED_SUSTAINABILITY

Specifications

Lid With Lid
Array 16 x 24
Packaging 15/Pk., 60/Cs.
Color Black, Clear Bottom
Material Polystyrene
No. of Wells 384
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
For Use With (Application) Tissue Culture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.