Learn More
LICOR GAR CellTag™ 700 Stain ICW Kit II
Supplier:  LICOR 92642092
Encompass_Preferred
GAR CellTag 700 Stain ICW Kits provide detection reagents for cell-based In-Cell Western™ Assays. This cost-effective normalization method makes quantification of the target protein more precise. CellTag 700 Stain accumulates in the nucleus and cytoplasm of permeabilized cells allowing for accurate normalization to cell number. This kit includes reagents to perform an In-Cell Western assay and CellTag 700 Stain to normalize well-to-well variations in cell number. CellTag 700 accumulates in the nucleus and cytoplasm of permeabilized cells allowing for accurate normalization to cell number. Kit Components include IRDye 800CW Goat anti-Rabbit Secondary Antibody (0.5 mg), Intercept (TBS) Blocking Buffer (1 x 500 mL), CellTag 700 Stain (2 x 10 nmol), and Kit Protocol
Catalog No. NC1726553