Learn More
Grainger CELLTREAT Non-Treated Plate, 96 Wells
Catalog No. NC2511985
Encompass_Preferred
The CELLTREAT Non-Treated 96-Well Plate is made of polystyrene for cell culture applications. It has a V-shaped bottom with an inner diameter of 6.4mm. The plate contains 96 wells arranged in 8 rows by 12 columns that are sterile and free of detectable DNase, RNase and pyrogens. It has a working volume of 300μl. The plates have a lid to prevent contamination and evaporation. They can be used for ELISA, tissue culture assays, cell-based assays and storage. The plates are non-treated for general cell attachment.
Catalog No. NC2511985 Supplier Grainger Supplier No. 38C826
Add to Cart
Add to Cart
The CELLTREAT Non-Treated 96-Well Plate is made of polystyrene for cell culture applications. It has a V-shaped bottom with an inner diameter of 6.4mm. The plate contains 96 wells arranged in 8 rows by 12 columns that are sterile and free of detectable DNase, RNase and pyrogens. It has a working volume of 300μl. The plates have a lid to prevent contamination and evaporation. They can be used for ELISA, tissue culture assays, cell-based assays and storage. The plates are non-treated for general cell attachment.