Learn More
CELLTREAT Scientific Products 9 Inch Polystyrene Plasteur Pasteur Pipet, Individually Wrapped Packed in Bags, Sterile
SDP
Catalog No. 50202134
Encompass_Preferred
The Plasteur Plastic Pasteur Pipet offers the aspirating precision of a glass pipet and the safety benefits of plastic. Fine tip is ideal for precision aspirating. No more broken glass: more durable than standard glass pipets. No damage during shipping and handling as compared to traditional glass pipets. Available sterile for convenience.
This item is not returnable. View return policy
Catalog No. 50-202-134 Supplier CELLTREAT Scientific Products Supplier No. 229280
The Plasteur Plastic Pasteur Pipet offers the aspirating precision of a glass pipet and the safety benefits of plastic. Fine tip is ideal for precision aspirating. No more broken glass: more durable than standard glass pipets. No damage during shipping and handling as compared to traditional glass pipets. Available sterile for convenience.