Learn More
CELLTREAT Scientific Products 25mL Serological Pipet, Individual Paper/Plastic Wrapper Packed in Bags, Sterile
SDP
Catalog No. 50202110
Encompass_Preferred
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification.
Catalog No. 50-202-110 Supplier CELLTREAT Scientific Products Supplier No. 229225B
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification.