Learn More
CELLTREAT Scientific Products 100mm x 20mm Tissue Culture Treated Dish w/Grip Ring, Sterile
SDP
Catalog No. 50202029
Encompass_Preferred
Cell Culture Dishes have thicker and more durable plastic than Petri Dishes. Cell culture treatment process provides consistent and even hydrophilic surface for improved cell attachment. Manufactured from medical grade virgin polystyrene. 35, 60, 70 and 100mm dishes available with a gripping feature for improved handling. Vented lids contain molded spacers to allow gas exchange, and stacking rings for increased stability.
Catalog No. 50-202-029 Supplier CELLTREAT Scientific Products Supplier No. 229620
Cell Culture Dishes have thicker and more durable plastic than Petri Dishes. Cell culture treatment process provides consistent and even hydrophilic surface for improved cell attachment. Manufactured from medical grade virgin polystyrene. 35, 60, 70 and 100mm dishes available with a gripping feature for improved handling. Vented lids contain molded spacers to allow gas exchange, and stacking rings for increased stability.