Learn More
CELLTREAT Scientific Products 6 Well Tissue Culture Plate, Sterile
SDP
Catalog No. 50202137
Encompass_Preferred
Tissue Culture Treated surface improves cell adhesion. Plates are labeled with alpha-numeric markings for easy identification of individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. Lids have a stacking feature for added stability when handling tall stacks of plates.
This item is not returnable. View return policy
Catalog No. 50-202-137 Supplier CELLTREAT Scientific Products Supplier No. 229106
Add to Cart
Add to Cart
Tissue Culture Treated surface improves cell adhesion. Plates are labeled with alpha-numeric markings for easy identification of individual wells. Each well has a raised rim which nests into the rings on the single direction lid to reduce evaporation and limit the risk of cross contamination. Lids have a stacking feature for added stability when handling tall stacks of plates.