Learn More
CELLTREAT Scientific Products 50mL Bio-Reaction Tube - Bag, Sterile
SDP
Catalog No. 50202012
Encompass_Preferred
Bio-reaction tubes offer a convenient and economical option for small scale, high density suspension cell culture. Ideal for biological reaction applications and free gas exchange. Aseptic conditions maintained through 0.22?m hydrophobic vented membrane. Useful in the screening and optimization processes of suspension cells. Reaction parameters can be optimized using small amounts of materials, maximizing the number of parallel experiments that can be performed simultaneously. Ideal for suspension cell culture, bacterial growth, seeding larger bioreactors, supporting start-up testing procedures for larger bioreactors, and lyophilization. Vent control labels for 50mL Bio-Reaction tube caps cover 1 to 5 of the cap vent openings to control the amount of gas exchange. Reaction parameters can be optimized using small amounts of materials, maximizing the number of parallel experiments that can be performed simultaneously.
Catalog No. 50-202-012 Supplier CELLTREAT Scientific Products Supplier No. 229475
Bio-reaction tubes offer a convenient and economical option for small scale, high density suspension cell culture. Ideal for biological reaction applications and free gas exchange. Aseptic conditions maintained through 0.22?m hydrophobic vented membrane. Useful in the screening and optimization processes of suspension cells. Reaction parameters can be optimized using small amounts of materials, maximizing the number of parallel experiments that can be performed simultaneously. Ideal for suspension cell culture, bacterial growth, seeding larger bioreactors, supporting start-up testing procedures for larger bioreactors, and lyophilization. Vent control labels for 50mL Bio-Reaction tube caps cover 1 to 5 of the cap vent openings to control the amount of gas exchange. Reaction parameters can be optimized using small amounts of materials, maximizing the number of parallel experiments that can be performed simultaneously.